missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ BCR™ Polyester Full-Finger Cleanroom Glove Liners
BIOPHARMA

Product Code. p-7230594 Shop All Berkshire Products
Click to view available options
Size:
Large
Regular
Unit Size:
10 pairs

Product Code. 12940975

Brand: Berkshire™ BGL3.20R

Only null left
Add to basket
Edge
Add to basket

Continuous filament knitted polyester construction is thin yet durable, while the full-finger design provides total protection for those with severe glove sensitivities.

  • Reduces perspiration and contact dermatitis commonly associated with wearing latex, urethane, PVC, or nitrile gloves
  • Full-finger design provides total protection for those with severe glove sensitivities
  • Can be worn alone as inspection gloves to prevent fingerprints on critical surfaces
  • 100% continuous filament knitted polyester is thin, yet durable
  • Ambidextrous design can be used on either hand
  • Gloves can be repeatedly laundered for reuse
  • Suitable for use in ISO Class Cleanrooms

Specifications

Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Material Polyester
Size Regular
Finger Style Full
For Use With (Application) Laboratories and Manufacturing Environments
Glove Cuff Style Knit
Hem Color White
Series BGL3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.