missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ BCR™ Polyester Half-Finger Cleanroom Glove Liners
BIOPHARMA

Product Code. p-7230588 Shop All Berkshire Products
Click to view available options
Size:
Large
Packaging Type:
Pack
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12415180

Brand: Berkshire™ BGL2.20L

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Continuous filament knitted polyester construction is thin yet durable, while the half-finger design permits tactility in addition to wicking moisture from the palms.

  • Reduces perspiration and contact dermatitis commonly associated with wearing latex, urethane, PVC, or nitrile gloves
  • Half-finger design permits tactility while also wicking moisture from the palms
  • 100% continuous filament knitted polyester is thin, yet durable
  • Ambidextrous design can be used on either hand
  • Gloves can be repeatedly laundered for reuse
  • Suitable for use in ISO Class Cleanrooms

Specifications

Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Material Polyester
Size Large
Finger Style Half
For Use With (Application) Laboratories and Manufacturing Environments
Glove Cuff Style Knit
Hem Color Blue
Packaging Type Pack
Series BGL2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.