missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BcuI (SpeI) (10 U/μL)

Cut at A^CTAGT sites with BcuI (SpeI) restriction enzyme, which performs best at 37°C in Tango buffer (Isoschizomers: AhlI, SpeI).

£62.00 - £247.00

Specifications

Concentration 10 U/μL
Methylation Sensitivity Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme BcuI (SpeI)
Compatible Buffer 10X Buffer Tango
Optimal Reaction Temperature 37°C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10589310
View Documents
Thermo Scientific™
ER1251
400 units
£62.00
400 units
Please to purchase this item. Need a web account? Register with us today!
10599310
View Documents
Thermo Scientific™
ER1252
2,000 units
£247.00
2000 units
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Highlights

  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Specifications

Specifications

10 U/μL
BcuI (SpeI)
37°C
Restriction Enzyme
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
10X Buffer Tango
No
Traditional Cloning
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ BcuI (SpeI) (10 U/μL)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.