missing translation for 'onlineSavingsMsg'
Learn More

BD 1063 dihydrochloride, Tocris Bioscience™

Product Code. 30001138
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective σ1 ligand, putative antagonist

σ1 receptor antagonist, approximately 50-fold selective over σ2 sites and ≥100-fold selective over opioid, PCP, muscarinic, dopamine, α1, α2, β adrenoceptor, 5-HT1 and 5-HT2 receptors. Prevents hyperlocomotion following cocaine administration in mice.

Chemical Identifiers

CAS 206996-13-6
Molecular Formula C13H20Cl4N2
Molecular Weight (g/mol) 346.117
InChI Key NXFDBTLQOARIMH-UHFFFAOYSA-N
Synonym bd 1063 dihydrochloride, bd1063 dhydrochloride, 1-2-3,4-dichlorophenyl ethyl-4-methylpiperazine dihydrochloride, bd 1063 dihydrochloride hplc, bd-1063 dhydrochloride;bd 1063 dhydrochloride, piperazine, 1-2-3,4-dichlorophenyl ethyl-4-methyl-, dihydrochloride
PubChem CID 11617161
IUPAC Name 1-[2-(3,4-dichlorophenyl)ethyl]-4-methylpiperazine;dihydrochloride
SMILES CN1CCN(CC1)CCC2=CC(=C(C=C2)Cl)Cl.Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 346.13
Percent Purity >99%
Chemical Name or Material BD 1063 dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.