Learn More
BD 7-AAD Staining Solution
Convenient, ready-to-use nucleic acid dye that can be used in place of propidium iodide (PI)
Brand: BD 559925
Additional Details : Weight : 0.02000kg
Alert:
7-AAD is a potential carcinogen. Avoid contact with skin and eyes. Wear suitable protective clothing, gloves, and eye/face protection.
Description
- 7-AAD: 7-Amino-Actinomycin D
- Format: Solution
- Can be used in conjunction with phycoerythrin (PE)- and fluorescein isothiocyanate (FITC)-labelled monoclonal antibodies in 2-color analysis
- Minimal spectral overlap between 7-AAD, PE and FITC fluorescence emissions
- Detection range: far red range of the spectrum
- Storage buffer: Aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
Flow cytometry
Specifications
Can be used in conjunction with Phycoerythrin (PE) -and Fluorescein Isothiocyanate (FITC) -labeled Monoclonal Antibodies in 2-color analysis | |
2 mL | |
7-AAD Staining Solution | |
4°C |
Research use only.