missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Annexin V: FITC Apoptosis Detection Kit I
Staining precedes the loss of membrane integrity which accompanies the latest stages of cell death resulting from either apoptotic or necrotic processes
Brand: BD 556547
323.16 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes: Annexin V-FITC, Propidium Iodide Staining Solution, Annexin V Binding Buffer
Description
- Characterized by certain morphologic features, including loss of plasma membrane asymmetry and attachment, condensation of the cytoplasm and nucleus, and internucleosomal cleavage of DNA
Specifications
1 Kit | |
4°C | |
FITC | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction