missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Annexin V-PE
Serves as a sensitive probe for flow cytometric analysis of cells that are undergoing apoptosis
£211.00 - £705.00
Specifications
Sample Volume | 5 μL |
---|---|
Content And Storage | 4°C |
For Use With (Application) | Flow Cytometry |
Product Code | Brand | Quantity | Storage Buffer | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Storage Buffer | Price | Quantity & Availability | ||||
12860460
|
BD
556422 |
100 Tests | Aqueous Buffered solution containing 0.2% BSA and 0.09% Sodium Azide |
£390.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10092374
|
BD
556421 |
200 Tests | Aqueous Buffered solution containing BSA and ≤0.09% Sodium Azide |
£705.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15840609
|
BD
560930 |
25 Tests | Aqueous Buffered solution containing BSA and ≤0.09% Sodium Azide |
£211.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Descrizione
- Format: PE
Flow Cytometry
Specifica
5 μL | |
4°C | |
Flow Cytometry |
Research Use Only