missing translation for 'onlineSavingsMsg'
Learn More

BD Annexin V-V450

Serves as a sensitive probe for flow cytometric analysis of cells that are undergoing apoptosis

Brand:  BD 560506

Additional Details : Weight : 0.01000kg

Product Code. 15890249

  • £414.00 / Each
Estimated Shipment: 22-08-2024
to see stock.

Description

Description

Identifies apoptosis at an earlier stage than assays based on nuclear changes such as DNA fragmentation.

  • Format: V450
  • Volume per Test: 5μL

Flow Cytometry

Specifications

Specifications

50 Tests
Aqueous Buffered solution containing BSA and ≤0.09% Sodium Azide
Flow Cytometry
5 μL
4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only