Learn More
BD Anti-rat Ig HRP Detection kit
Designed for immunohistochemical staining of acetone-fixed frozen sections, formalin- or zinc-fixed paraffin sections, and cytospins/smears
Brand:  BD 551013
Additional Details : Weight : 0.65000kg
Product Code. 15816478
  • £660.00 / 1 set
Estimated Shipment: 23-08-2024
to see stock.
Add to basket
Includes: Biotinylated goat anti-rat Ig, Antibody Diluent Buffer, Streptavidin-Horseradish peroxidase (SAv-HRP), DAB Buffer, and DAB Chromogen
Description
Description
  • Used with any purified primary antibody
  • Provides maximum reactivity
  • Contains protein stabilizers to enhance antibody reactivity
  • Blocking agents reduce non-specific staining
Immunohistochemistry
Specifications
Specifications
1 Kit
4°C
Immunohistochemistry, Immunohistochemical Staining