missing translation for 'onlineSavingsMsg'
Learn More

BD Bacto™ Dehydrated Agar

Product Code. 11758223
Click to view available options
Quantity:
100 g
454 g
2000 g
Packaging Type:
Bottle
Container
Unit Size:
100g
2kg
454g
This item is not returnable. View return policy

Product Code. 11758223

Brand: BD 214030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purified solidifying agent. BD Difco™ Bacto™ Dehydrated Agar is purified to remove extraneous matter.

Purified solidifying agent. BD Difco™ Bacto™ Dehydrated Agar is purified to remove extraneous matter.

  • Extraneous matter, pigmented portions and salts are reduced to minimum
  • Fine, light colored granules
  • Convenient for weighing and handling

Determines motility and growth of anaerobes and microaerophiles

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Concentration 1.5%
Additive Agar
Form Dehydrated
For Use With (Application) Determination of motility and the growth of anaerobes and microaerophiles
Packaging Type Container
Quantity 2000 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.