missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Bacto™ Dehydrated Agar
Purified solidifying agent. BD Difco™ Bacto™ Dehydrated Agar is purified to remove extraneous matter.
£130.03 - £2055.00
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
---|---|
Concentration | 1.5% |
Additive | Agar |
Form | Dehydrated |
For Use With (Application) | Determination of motility and the growth of anaerobes and microaerophiles |
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
11768223
|
BD
214050 |
Bottle | 100 g |
£130.03
100g |
In Stock
Log in to see stock. |
||||
10455513
|
BD
214010 |
Bottle | 454 g |
£940.15
454g |
In Stock
Log in to see stock. |
||||
11758223
|
BD
214030 |
Container | 2000 g |
£2055.00
2kg |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
Description
Purified solidifying agent. BD Difco™ Bacto™ Dehydrated Agar is purified to remove extraneous matter.
- Extraneous matter, pigmented portions and salts are reduced to minimum
- Fine, light colored granules
- Convenient for weighing and handling
Determines motility and growth of anaerobes and microaerophiles
Specifications
ISO/DIS, 11014 | |
Agar | |
Determination of motility and the growth of anaerobes and microaerophiles |
1.5% | |
Dehydrated |