Learn More
BD BBL™ AFB Slides
For evaluating and controlling the quality of acid-fast and fluorescent stain reagents and staining techniques for the detection of acid-fast bacilli (AFB)
Brand:  BD 231391
Product Code. 11722964
  • £889.00 / Pack of 50
Explore more special offers
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD BBL™ AFB Slides > AFB QC Control Slides
For in vitro diagnostic use.