Learn More
BD BBL™ Chopped Meat Carbohydrate Broth, Pre-Reduced II (Hungate Cap)
General-purpose medium for enrichment and cultivation of anaerobic microorganisms, particularly obligate anaerobes
Brand: BD 297307
Additional Details : Weight : 0.01980kg
Requires : Ancillary culture media, reagents, quality control organisms and laboratory equipment
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
CE, CMPH2 | |
2 to 25°C | |
United States | |
Enrichment and cultivation of anaerobic microorganisms, obligate anaerobes | |
10/Pk. |
Beige | |
Liquid | |
Chopped meat Pellets 10.2g/L, Pancreatic digest of Casein 30g/L, Yeast extract 5g/L, Glucose 4g/L, Dipotassium Phosphate 5g/L, Cellobiose 1g/L, Maltose 1g/L, Starch 1g/L, L-Cysteine HCl 0.5g/L, Resazurin 0.001g/L, Vitamin K1 1mg/L, Hemin 5mg/L | |
Tube |
For in vitro diagnostic use