missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Coagulase Plasma, Rabbit, with Citrate

Reconstitutes for use in the Direct Tube Method. BD™ BBL™ Coagulase Plasma, Rabbit, with Citrate is a lyophilized reagent to qualitatively identify and determine the pathogenicity of Staphylococcus aureus.

Brand:  BD 240661

 View more versions of this product

Product Code. 11738852

  • £366.00 / Pack of 10
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Identification of staphylococci is based on microscopic examination, colonial morphology and cultural and biochemical characteristics. Staphylococci associated with acute infection can clot plasma. The most widely used and generally accepted criterion for identification of these pathogenic organisms is based on the presence of the enzyme coagulase.

Staphylococcus aureus produces two types of coagulase, free and bound. Free coagulase is an extracellular enzyme produced when the organism is cultured in broth. Bound coagulase, also known as the clumping factor, remains attached to the cell wall of the organism. In the direct tube test, free coagulase liberated from the cell acts on prothrombin in the coagulase plasma to give a thrombin-like product. This product then acts on fibrinogen to form a fibrin clot.

  • Available in 10-tube packs that reconstitute to 3ml or 15ml for use in the Direct Tube Method
  • Store reconstituted plasma at 2–8°C for up to 14 day, or aliquot and freeze promptly for up to 30 days
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rabbit
2°C to 8°C
Coagulase Rabbit Plasma
Qualitative procedures for the identification of Staphylococcus aureus
Light brown
Lyophilized, Liquid
Lyophilized rabbit plasma with 0.85% sodium citrate and 0.85% sodium chloride
10 x 15 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.