Learn More
BD BBL™ Dehydrated Culture Media: Soy Agar with Lecithin and Polysorbate 80
Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms
£434.00
Specifications
Form | Dehydrated |
---|---|
Media Type | Soy Agar with Lecithin and Polysorbate 80 |
Product Type | Agar |
Description
Detection of microorganisms on surfaces sanitized with quaternary ammonium compounds
Certifications
CE, CCAM
Specifications
Dehydrated | |
Soy Agar with Lecithin and Polysorbate 80 | |
Agar |
Safety and Handling
missing translation for 'shelfLife' : 1460 Days
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BD BBL™ Dehydrated Culture Media: Soy Agar with Lecithin and Polysorbate 80