missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Penicillinase Concentrate
For cultivation of heterotrophic microorganisms
£178.00 - £401.00
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
---|---|
Additive | Penicillinase |
Form | Liquid |
For Use With (Application) | Formulation of culture media for the cultivation of heterotrophic microorganisms |
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
10615886
|
BD
211898 |
Tube | 20 mL |
£401.00
200mL |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
10369787
|
BD
211899 |
Bottle | 100 mL |
£178.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
Specifications
ISO/DIS, 11014 | |
Penicillinase | |
Liquid | |
Formulation of culture media for the cultivation of heterotrophic microorganisms |