missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Peptone

Bacto Peptone is an animal-origin (AO) enzymatic digest of bovine and porcine animal proteins.

Brand:  Gibco™ 211830

 View more versions of this product

Product Code. 16229761

  • £2070.00 / 10kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Bacto Peptone is an animal-origin enzymatic digest of bovine and porcine animal proteins. The nutritive value of Bacto Peptone is largely dependent on a combination of a negligible quantity of proteases, high concentration of complex constituents, and a unique amino acid content that makes this peptone a strong nitrogen source. Customers have successfully used this supplement with a variety of cell types including Saccharomyces cerevisiae, Escherichia coli, Pasteurella multocida, Vibrio cholerae, and porcine kidney cells to produce biotherapeutics and vaccines for animal health applications.

Features:

  • A strong nitrogen source
  • Unique amino acid content
  • High concentration of complex constituents

Applications of Bacto Peptone include:

  • Vaccines for animal health applications
  • Production of biotherapeutics
  • Mammalian cell culture
  • Microbial fermentation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LUO, MUO
Bacteria
Bacterial Media
Formulation of culture media for the cultivation of heterotrophic microorganisms
10 kg
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Powder
Pail
Bacto™
Animal origin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers