missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Peptone

Bacto Peptone is an animal-origin (AO) enzymatic digest of bovine and porcine animal proteins.

Brand:  Gibco™ 211677

 View more versions of this product

Product Code. 16299741

  • £128.00 / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Bacto Peptone is an animal-origin enzymatic digest of bovine and porcine animal proteins. The nutritive value of Bacto Peptone is largely dependent on a combination of a negligible quantity of proteases, high concentration of complex constituents, and a unique amino acid content that makes this peptone a strong nitrogen source. Customers have successfully used this supplement with a variety of cell types including Saccharomyces cerevisiae, Escherichia coli, Pasteurella multocida, Vibrio cholerae, and porcine kidney cells to produce biotherapeutics and vaccines for animal health applications.

Features:

  • A strong nitrogen source
  • Unique amino acid content
  • High concentration of complex constituents

Applications of Bacto Peptone include:

  • Vaccines for animal health applications
  • Production of biotherapeutics
  • Mammalian cell culture
  • Microbial fermentation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LUO, MUO
Bacteria
Powder
Bottle
Bacto™
Animal origin
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Mammalian, Insect, Microbial
Bacterial Media
Formulation of culture media for the cultivation of heterotrophic microorganisms
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Bacto™ Peptone > 500 g, Bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.