missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Proteose Peptone No. 3

Enzymatic digest of porcine protein

Brand:  Gibco™ 211693

Additional Details : Weight : 0.52200kg

Product Code. 16249751

  • £324.00 / 500g
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Bacto Proteose Peptone No. 3 is a member of the Bacto Proteose Peptones, which are enzymatic digests of protein
  • Studies show that no single peptone is the most suitable nitrogen source for every microbiological application
  • These findings led to the development of Bacto Proteose Peptone, Bacto Proteose Peptone No. 2, and Bacto Proteose Peptone No. 3. Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone
  • Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Specifications

Specifications

LUO, MUO
Mammalian, Insect, Microbial
Bacterial Media
Enzymatic digest of Protein
500 g
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Powder
Bottle
Bacto™
Animal Origin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers