missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Skim Milk
Differentiate organisms based on coagulation and proteolysis of casein. BBL™/Difco™ Media Additives/Ingredients/Reagents: Skim Milk can be added to media.
Brand: BD 232100
Code : TP
Additional Details : Weight : 0.53100kg
Description
- Differentiation of organisms based on coagulation and proteolysis of casein
Specifications
OSHA, HCS | |
Skim Milk | |
Preparing microbiological culture media | |
500 g |
<30°C | |
Dehydrated | |
Bottle |