Learn More
BD BBL™/Difco™ TB Brilliant Green K
For staining mycobacteria by the Kinyoun (cold) and Ziehl-Neelsen (hot) acid-fast procedures
Brand:  BD 212523
Product Code. 11768193
  • £116.00 / 1 L
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Stains smears prepared from specimens and cultures suspected of containing mycobacteria for early presumptive diagnosis of mycobacterial infection and characterization of bacterial isolates.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TB Brilliant Green K
For staining mycobacteria by the Kinyoun (cold) and Ziehl-Neelsen (hot) acid-fast procedures
Bottle
4 × 250mL
15° to 30°C
Brilliant Green K
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For in vitro Diagnostic Use.