Learn More
BD BBL™ Hemo ID Quad Plate, with Growth Factors
Separates various growth factors by media type. Isolates, differentiates and identifies Haemophilus species based on the X- and V-factor growth requirements and the hemolysis of horse blood.
Brand:  BD 297890
Product Code. 16544170
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Facilitates identification and eliminates the difficulties associated with inspecting and interpreting undivided colonies
  • After 24 hours of incubation, the Quad Plate will indicate the presence or absence of growth in specific quadrants, depending on X- and V-factor requirements for the various Haemophilus species
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hemo ID Quad Plate
Growth factors
10 Pack