missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ IsoVitaleX™ Enrichment
BD BBL™ IsoVitaleX™ Enrichment is a chemically defined supplement used as an additive to media for cultivation of nutritionally fastidious microorganisms.
£372.00
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
---|---|
Content And Storage | 2°C to 8°C |
Additive | IsoVitaleX™ Enrichment |
Form | Lyophilized |
For Use With (Application) | Dehydrated culture media |
Description
- GC Agar supplemented with hemoglobin and BBL IsoVitaleX Enrichment (in place of yeast concentrate) provides an improved medium for the isolation of microorganisms
- Enrichment provides V factor (nicotinamide adenine dinucleotide, NAD) for Haemophilus species
- Also provides vitamins, amino acids, coenzymes, dextrose, ferric ions and other factors which improve the growth of pathogenic Neisseria
Specifications
ISO/DIS, 11014 | |
IsoVitaleX™ Enrichment | |
Dehydrated culture media |
2°C to 8°C | |
Lyophilized | |
Vial |
Safety and Handling
missing translation for 'shelfLife' : 1095 Days
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Isovitalex Enrichment
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P363-Wash contaminated clothing before reuse.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Hydrochloric Acid