Learn More
BD BBL™ Medium Herrold's Egg Yolk Agar: With Mycobactin J
Selective isolation and differentiation of Mycobacterium paratuberculosis
Brand: BD 222233
Additional Details : Weight : 0.10000kg
Description
- Available with amphotericin, nalidixic Acid, vancomycin
- Features “Slant Surface”, a widely used format that gives enough surface area for growth and visualization
Specifications
ISO/DIS, 11014 | |
Liquid | |
Herrold's Egg Yolk Agar with Amphotericin, Nalidixic Acid, Vancomycin, with Mycobactin J | |
Slant |
Light Yellow | |
United States | |
Selective isolation and differentiation of Mycobacterium paratuberculosis | |
100/Box |