missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Mycoflask™ Prepared Tubed Media: BHI broth with 6.5% NaCl
For cultivation of fastidious and nonfastidious microorganisms from a vriety of clinical and nonclinical materials
Marca: BD 221785
Detalles adicionales : Weight : 0.01800kg
Descripción
- For cultures requiring prolonged incubation
- Contains 6.5% sodium chloride
Especificaciones
ISO/DIS, 11014, AOAC, BAM, CCAM, CLSI, CMPH2, COMPF, EPA, ISO, MCM9, SMD, SMWW, USDA | |
2 to 25°C | |
United States | |
Brain heart infusion from (solids) 6g/L, Peptic digest of Animal Tissue 6g/L, Sodium Chloride 5g/L, Dextrose 3g/L, Pancreatic digest of Gelatin 14.5g/L, Disodium Phosphate 2.5g/L | |
Tube |
Beige | |
Liquid | |
Brain heart infusion broth | |
General purpose liquid medium used in the cultivation of fastidious and non-fastidious microorganisms | |
0.5 mL Tubes, 10 Pack |