Learn More
BD BBL™ Mycoflask™ Prepared Tubed Media: Bile Esculin Agar Slants
Used to differentiate enterococci and the Streptococcus bovis group from other streptococci
Brand: BD 221410
Additional Details : Weight : 0.02070kg
Description
- For cultures requiring prolonged incubation
Specifications
CE, COMPF | |
2°C to 8°C | |
United States | |
Pancreatic digest of Gelatin 5g/L, Beef extract 3g/L, Oxgall 20g/L, Ferric Citrate 0.5g/L, Esculin 1g/L, Agar 14g/L | |
Tube |
Beige | |
Solid | |
Bile Esculin Agar | |
Isolation and cultivation of pathogenic and nonpathogenic fungi | |
100/Pk. |
Safety and Handling
- BD BBL Bile Esculin Agar Slants , 100 pack 6.8 Ml
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : bile extract