missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Mycoflask™ Sabouraud Dextrose Agar Prepared Media Slants with Chloramphenicol
Used in qualitative procedures for cultivation of dermatophytes
Brand: BD 221825
Additional Details : Weight : 0.02950kg
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
ISO/DIS, 11014, MCM9 | |
2°C to 8°C | |
United States | |
Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Agar 15g/L | |
Tube with Tight Cap |
Beige | |
Liquid | |
Sabouraud Dextrose Agar | |
Cultivation of dermatophytes | |
100 Pack |
For in vitro diagnostic use.