missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Mycoflask™ Sabouraud Liquid Broth Prepared Media, Modified
Used for the cultivation of yeast and molds
Brand: BD 221014
Additional Details : Weight : 0.80000kg
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
ISO/DIS, 11014, AOAC, USP | |
2 to 25°C | |
United States | |
Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 20g/L | |
Tube with Tight Cap |
Beige | |
Liquid | |
Sabouraud Liquid Broth | |
Cultivation of Yeast and molds | |
10/Pk. |
For in vitro diagnostic use.