missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Sensi-Disc™ Antibiotic Susceptibility Test Discs Accessories: Sensi-Disc Dispenser
£1000.00
Specifications
For Use With (Equipment) | Sensi-Disc™ antibiotic susceptibility test disc |
---|---|
Includes | Storage canister, reusable indicator desiccant container |
Quantity | Each |
Description
- Self-tamping, complete with storage canister and reusable indicator desiccant container
- Choose from 6-place or 8-place
Specifications
Sensi-Disc™ antibiotic susceptibility test disc | |
Storage canister, reusable indicator desiccant container | |
Each |