Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Ciprofloxacin31
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
£44.15
Specifications
Antimicrobial Agents | Ciprofloxacin31 |
---|---|
Concentration | 5μg |
Packaging Type | Disc |
Description
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Ciprofloxacin31 | |
5μg | |
Disc |
For in vitro diagnostic use