missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Staphyloslide™ Latex Test Kit

Provides a rapid platform for the identification of Staphylococcal isolates, particularly, Staphylococcus aureus

£226.00

Specifications

Content And Storage 2°C to 8°C Do not freeze
Description BBL Staphyloslide Latex Test Kit
Detectable Analytes S. aureus
For Use With (Application) Staphylococcus aureus Testing
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15410937
View Documents
BD
240915
100 Tests
£226.00
1 set
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The BD BBL Staphyloslide Latex Test Kit utilizes blue polystyrene latex particles that have been sensitized with fibrinogen and IgG. When Staphylococcal colonies which possess bound coagulase (clumping factor) and/or protein A are mixed with the latex reagent, the latex particles will agglutinate strongly within 20 seconds.

Specifications

Specifications

2°C to 8°C Do not freeze
BBL Staphyloslide Latex Test Kit
S. aureus
Staphylococcus aureus Testing
Videos
SDS
Documents

Documents

Product Certifications