missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biotin Human NK Cell Enrichment Set - DM
Negative selection of Natural Killer (NK) cells from peripheral blood
Brand: BD 557987
Additional Details : Weight : 0.02000kg
Includes: CD3, CD19, CD36, CD41a, CD66b, CD123, CD235a, IgE biotinylated antibody cocktail and BD IMag SAv particles
Description
Set consists of Biotinylated Human NK Cell Enrichment Cocktail and BD IMag™ Streptavidin Particles Plus
- Reactivity: Human
- Format: BD IMag-DM, Biotin
Magnetic Labeling
Specifications
Biotin | |
1 x 109 Cells |
Research Use Only