missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Canine C-reactive protein (CRP) ELISA Kit
CRP quantitation for determination of inflammatory conditions
Brand: BD 557826
Additional Details : Weight : 0.65000kg
Includes: Capture antibody coated microplate, detection antibody/enzyme conjugate (100x), standard (10x), wash buffer, TMB substrate, stop solution
Description
For detection and quantitation of canine CRP in canine serum
- Reactivity: Dog
- Utilizes an antibody specific for canine CRP coated on a 96-well plate
Enzyme-linked immunosorbent assay
Specifications
Dog | |
1 Plate | |
2°C to 8°C | |
Canine C-reactive Protein [CRP] ELISA kit |
Capture Antibody coated microplate, detection Antibody/Enzyme Conjugate (100X), standard (10X), Wash Buffer, TMB Substrate, stop solution | |
Serum | |
Detection and quantitation of Canine CRP in Canine Serum, ELISA |
Research Use Only