Learn More
BD Caspase-3 Assay Kit
Designed to measure caspase-3 or DEVD-cleaving activity an marker of cells undergoing apoptosis.
Brand:  BD 556485
Additional Details : Weight : 1.02000kg
Product Code. 15818718
  • £575.00 / 1 set
Estimated Shipment: 22-08-2024
to see stock.
Includes: PBS Buffer, Cell Lysis Buffer, HEPES Buffer, Caspase-3 Fluorogenic Substrate, Caspase-3 Inhibitor
Description
Description
  • Used to determine if certain apoptosis pathways involve caspase-3 activation
Specifications
Specifications
1 Kit
4°C
Determine if certain apoptosis pathways involve Caspase-3 activation