missing translation for 'onlineSavingsMsg'
Learn More

BD Cell Enrichment Cocktail

Product Code. 15819458
Click to view available options
Product Type:
Memory CD4+ T Cell Cocktail
Naive CD4+ T Cell Cocktail
Naive CD8+ T Cell Cocktail
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15819458

Brand: BD 558521

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Human Memory CD4+ T Cell Enrichment Cocktail, 5.0mL:

  • Biotin Mouse Anti-human CD8, clone SK1
  • Biotin Mouse Anti-human CD11b, clone ICRF44
  • Biotin Mouse Anti-human CD16, clone 3G8
  • Biotin Mouse Anti-human CD19, clone HIB19
  • Biotin Mouse Anti-human CD36, clone CB38
  • Biotin Mouse Anti-human CD41a, clone HIP8
  • Biotin Mouse Anti-human CD45RA, clone HI100
  • Biotin Mouse Anti-human CD56, clone B159
  • Biotin Mouse Anti-human CD123, clone 9F5
  • Biotin Mouse Anti-human CD235a (Glycophorin A), clone GA-R2 (HIR2)
  • Biotin Mouse Anti-human γδ TCR, clone B1
  • In aqueous buffered solution containing 0.09% Sodium Azide

BD IMag Streptavidin Particles Plus - DM, 7.5mL:

  • In aqueous buffered solution containing BSA™ and 0.09% Sodium Azide

For negative selection of untouched memory CD4+ helper T cells from peripheral blood

Contains biotinylated monoclonal antibodies that recognize antigens expressed on naive T cells, CD8+ T cells, B cells, NK cells, Γ/δ T cells, monocytes, DCs, granulocytes, platelets, and erythroid cells
Negative selection is performed to enrich for unlabeled memory cells. Labeled cells migrate towards magnet (positive fraction), leaving the unlabeled cells in suspension so they can be drawn off and retained (enriched fraction)

  • Contains biotinylated monoclonal antibodies that recognize antigens expressed on naive T cells, CD8+ T cells, B cells, NK cells, γ/δ T cells, monocytes, DCs, granulocytes, platelets, and erythroid cells
  • Avoids inadvertent activation of enriched memory helper T cells by using reagents that do not directly bind to those T cells
  • Simultaneously stains erythrocytes, platelets, and most leukocytes

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage In aqueous buffered solution containing BSA™ 0.09% sodium azide at 4°C
For Use With (Application) For negative selection of untouched memory CD4+ helper T cells from peripheral blood, magnetic cell separation
For Use With (Equipment) Flow cytometer
Includes Biotin mouse Anti-Human CD8, clone SK1, CD11b, clone ICRF44, CD16, clone 3G8, CD19, clone HIB19, CD36, clone CB38, CD41a, clone HIP8, CD45RA, clone HI100, CD56, clone B159, CD123, clone 9F5, CD235a (Glycophorin A), clone GA-R2 (HIR2), gd TCR, clone B1
Product Type Naive CD4+ T Cell Cocktail
Quantity 5 mL

Research use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.