Learn More
BD Cell Viability Solution
Ready-to-use solution of the nucleic acid dye, 7-Amino-actinomycin D (7-AAD) that can be used in place of propidium iodide (PI) for the exclusion of nonviable cells in flow cytometric assays
Brand:  BD 555815
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 15868458
  • £475.00 / Each
Estimated Shipment: 18-10-2024
to see stock.
Description
Description
  • Used as a viability probe for methods of dead cell exclusion, based on a light scatter and uptake of 7-AAD as detected in FL3
  • Does not require dilution
Flow Cytometry
Specifications
Specifications
4°C
Flow Cytometry
500 Tests