missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD CompBead Plus Anti-Rat Ig, κ/Negative Control Compensation Plus (7.5μm) Particles Set
Provides two types of particles, CompBead Plus Anti-Rat Ig, κ particles, which binds any rat κ light chain-bearing immunoglobulin, and CompBead Plus Negative Control (BSA) particles, which has no binding capacity
Brand: BD 560499
Additional Details : Weight : 0.02000kg
Includes:
- Negative Control (PBS with 1% BSA)
- Anti-Rat Ig, κ
Description
- Mixed together with fluorochrome-conjugated rat antibody, CompBead Plus provides distinct positive and negative (background fluorescence) stained populations which can be used to set compensation levels manually or using instrument set-up software
Specifications
Flow Cytometry | |
1 Each | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |