Learn More
BD Cytofix/Cytoperm™ Fixation and Permeabilization Solution
1X solution can be used for the simultaneous fixation and permeabilization of cells prior to intracellular cytokine staining
Brand: BD 554722
Additional Details : Weight : 0.18000kg
Description
Specifications
Can be used for the simultaneous fixation and permeabilization of cells prior to intracellular Cyto kine staining | |
Store undiluted at 4°C | |
125mL |
7.2 | |
Fixation and permeabilization solution | |
Liquid |
Safety and Handling
- BD Cytofix/Cytoperm Fixation and Permeabilization Solution
Signal Word
- Danger
Hazard Category
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
Hazard Statement
- H317-May cause an allergic skin reaction.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P501-Dispose of contents/container to an appropriate treatment and disposal facility in accordancewith applicable laws and regulations, a.
Supplemental information
- MIXTURE LIST-Contains : formaldehyd e...%