Learn More
BD Cytometric Bead Array (CBA) Functional Bead Conjugation Buffer Set
Used to covalently link watersoluble proteins to surface of color-coded functional beads
Brand: BD 558556
Additional Details : Weight : 0.36000kg
Alert:
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.
Includes: Coupling buffer and storage buffer
Description
- Contains supporting reagents for BD™ CBA Functional Bead system
- Simple conjugation procedure completed within 4 hours
- Conjugation reaction requires less then 100μg of protein at a concentration of 1mg/mL
- Size: 15 tests
Specifications
RUO | |
Flow Cytometry | |
BD™ CBA functional bead system | |
15 |
4°C | |
Functional Bead Conjugation Buffer Set | |
Coupling buffer and storage buffer and supporting reagents |