Learn More
BD Cytometric Bead Array (CBA) Functional Beads
Color-coded 7.5μm polystyrene beads that can be classified using a flow cytometer
Brand: BD 558646
Additional Details : Weight : 0.01000kg
Description
The Functional Beads can be used in conjunction with the BD CBA Functional Bead Conjugation Buffer Set (Mfr. No. 558556) to covalently link water-soluble proteins to the surface of the beads via a sulfo-SMCC chemistry. The beads, once linked to a protein, can be used as a capture matrix for analytes of interest.
Specifications
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide | |
Flow Cytometry | |
Functional Bead | |
6000 | |
7.5 μm |
4°C | |
Functional Bead (B9) | |
Polystyrene | |
Flow cytometer | |
0.9 mL |