missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Functional Beads

Color-coded 7.5μm polystyrene beads that can be classified using a flow cytometer

Brand:  BD 560031

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15889868

  • £1105.00 / Each
Estimated Shipment: 06-08-2024
to see stock.

Description

Description

The Functional Beads can be used in conjunction with the BD CBA Functional Bead Conjugation Buffer Set (Mfr. No. 558556) to covalently link water-soluble proteins to the surface of the beads via a sulfo-SMCC chemistry. The beads, once linked to a protein, can be used as a capture matrix for analytes of interest.

Specifications

Specifications

29 CFR 1910.1200, 91/155/EEC, 93/112/EC, RUO
Aqueous buffered solution containing ≤0.09% sodium azide
Clear
Polystyrene
Liquid
6000
7.5 μm
4°C
Functional Bead (A5)
Flow Cytometry
Functional Bead
Flow cytometer
0.9 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers