missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Human Inflammatory Cytokine Kit Standard controls
Have been tested to assure function as standards in CBA assays
£1515.00 - £1720.00
Specifications
Cross Reactivity | Human |
---|---|
Content And Storage | 4°C |
Form | Lyophilized |
For Use With (Application) | Flow Cytometry |
Product Code | Brand | Product Type | Includes | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Includes | Quantity | Price | Quantity & Availability | |||
15886898
|
BD
552932 |
Human inflammatory Cytokine lyophilized standard | Mixture of recombinant Human Interleukin (IL)-8, IL-1beta, IL-6, IL-10, Tumor Necrosis Factor (TNF) and IL-12p70 | 0.2 mL |
£1515.00
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||
15811219
|
BD
561666 |
Human Th1/Th2/Th17 Cytokine standards | Mixture of recombinant Human interleukin (IL)-2, IL-4, IL-6, IL-10, IL-17A, interferon-g (IFN-g) and Tumor Necrosis Factor (TNF) | 1 Vial |
£1720.00
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||
Description
- Lyophilized in aqueous buffered solution containing BSA and ProClin™ 150
- Should be reconstituted and serially diluted before use
Human Inflammatory Cytokine Standards
- Contain mixture of recombinant human Interleukin (IL)-8, IL-1β, IL-6, IL-10, Tumor Necrosis Factor (TNF), and IL-12p70
- Use with BD™ Cytometric Bead Array (CBA) Human Inflammatory Cytokine Kit
- Size: 0.2mL
- Contain mixture of recombinant human Interleukin (IL)-2, IL-4, IL-6, IL-10, IL-17A, Interferon-γ (IFN-γ), and Tumor Necrosis Factor (TNF)
- Use with BD™ Cytometric Bead Array (CBA) Human Th1/Th2/TH17 Cytokine Kit
- Size: 1 vial
Flow Cytometry
Specifications
Human | |
4°C | |
Lyophilized | |
Flow Cytometry |