missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Mouse IL-1α; Flex Set, Bead E4

Determines mouse reactivity

Brand:  BD 560157

Additional Details : Weight : 0.99000kg


Product Code. 10208893

  • £526.00 / Each
Estimated Shipment: 26-07-2024
to see stock.
Add to basket

Includes: Mouse IL-1α; Standard; Mouse IL-1α; PE Detection Reagent; Mouse IL-1α; Capture Bead E4

Description

Description

The BD CBA Mouse IL-1α Flex Set is a bead-based immunoassay capable of measuring mouse interleukin-1α (IL-1α) in serum and cell culture supernatant samples. Mouse reactivity was determined by testing samples with the BD CBA Mouse IL-1α Flex Set.

  • Reactivity: QC Tested: Mouse
  • Bead Position: E4
  • Limit of Detection: 0.8pg/mL

Specifications

Specifications

IL-1alpha
Mouse IL-1a, standard, Mouse IL-1a, PE detection reagent, Mouse IL-1a, capture bead E4
4°C
Mouse IL-1alpha flex set (bead E4)
Mouse
100 Tests
Flow Cytometry
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BD Cytometric Bead Array (CBA) Mouse IL-1

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : 5-chloro-2-methyl-4-isothiazolin-3-one,2-methyl-2H-isothiazol-3-one

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research use only