missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Mouse/Rat Soluble Protein Master Buffer Kit, 500 tests

Employs particles with discrete fluorescence intensities to detect soluble analytes at very low concentrations

Brand:  BD 558267

Additional Details : Weight : 1.90100kg

Product Code. 15859268

  • £1680.00 / 1 set
Estimated Shipment: 18-10-2024
to see stock.

Includes: Assay Diluent, Capture Bead Diluent, Detection Reagent Diluent, Wash Buffer, Instrument Setup Beads (A1, A9, F1, F9), PE Instrument Setup Bead F1, PE Positive Control Detector

Description

Description

This kit contains all supporting reagents necessary to perform an assay using BD™ CBA Mouse or Rat Soluble Protein Flex Set. Buffers and instrument setup reagents are optimized for analysis of analytes in tissue culture supernatants, plasma and serum samples.

  • Analyte working assay range: 10 to 2500pg/mL (mouse), 40 to 10000pg/mL (rat)
  • Should not be used with any non-Mouse or Rat Soluble Protein Flex Sets
  • Available in pack sizes of 100 and 500 tests

Flow cytometry

Specifications

Specifications

2°C to 8°C
Flow cytometer
1 bottle 150 mL assay diluent, 1 bottle 30 mL capture bead diluent, 1 bottle 30 mL detection reagent diluent, 1 bottle 650 mL wash buffer, 1 vial 0.25 mL instrument setup bead A1, 1 vial 0.25 mL instrument setup bead A9, 1 vial 1.0 mL instrument setup bead F1
For analysis of analytes in tissue culture supernatants, plasma and serum samples
500
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers