missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array Flex Set Standard
Used as standard control
Brand: BD 558474
This item is not returnable.
View return policy
Description
This product contains lyophilized recombinant human Interferon-λ (IFN-λ)
- Storage Buffer: Lyophilized in aqueous buffered solution containing BSA and ProClin™ 150
Flow cytometry
Specifications
4°C | |
Lyophilized | |
100 | |
RUO |
Mouse IL-10 Flex Set Standard | |
Lyophilized recombinant Human Interferon-l (IFN-l) | |
0.2 mL | |
Cytometric Bead Array (CBA) Standard |