missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array Human RANTES Standard

Lyophilized human RANTES

Brand:  BD 558470

Product Code. 15829418

  • £168.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
  • Protein concentration of 25,000pg/ml (when reconstituted in 0.4mL Assay Diluent)
  • Regulatory status: RUO
  • Recombinant

Flow cytometry, controls

Order Info

100 tests.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aqueous Buffered Solution containing BSA and ProClin™ 150
100 Tests
Flow cytometry
25000pg/mL
0.2mL
Lyophilized
Technical data sheet
Cytometric Ab Control Standards
RUO
4°C
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers