Learn More
Cytometric Bead Array Human Th1/Th2/Th17 Kit, BD
Optimized for analysis of physiologically relevant concentrations (pg/mL levels) of specific cytokine proteins in tissue culture supernatants, EDTA plasma, and serum samples
Brand: BD Biosciences 560484
Additional Details : Weight : 0.56000kg
Includes: IL-2, IL-4, IL-6, IL-10, TNF, IFN-γ, IL-17A
Description
Detection reagent provided in the kit is a mixture of phycoerythrin (PE)–conjugated antibodies, which provides a fluorescent signal in proportion to the amount of bound analyte.
- Can be used to measure Interleukin-2 (IL-2), Interleukin-4 (IL-4), Interleukin-6 (IL-6), Interleukin-10 (IL-10), Tumor Necrosis Factor (TNF), Interferon-γ (IFN-γ), and Interleukin-17A (IL-17A) protein levels in a single sample
- Provides sufficient reagents for 80 tests
For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications
2° to 8°C | |
80.0 | |
1 vial 0.8mL human IL-2 capture beads, 1 vial 0.8mL human IL-4 capture beads, 1 vial 0.8mL human IL-6 capture beads, 1 vial 0.8mL human IL-10 capture beads, 1 vial 0.8mL human TNF capture beads, 1 vial 0.8mL human IFN capture beads | |
RUO |
Safety and Handling
- BD Cytometric Bead Array (CBA) Human Th1/Th2/Th17 Kit
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 5-chloro-2-methyl-4-isothiazolin-3-one