missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array Mouse Th1/Th2 Cytokine Standard

Product Code. 15866908
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15866908

Brand: BD 552967

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for use as a standard control with the Cytometric Bead Array (CBA) Mouse Th1/Th2 Cytokine Kit

  • Contains a mixture of individually lyophilized recombinant proteins made up of recombinant mouse IL-2, IL-4, IL-5, IFN-γ, and TNF
  • Lyophilized and should be reconstituted and serially diluted before mixing with CBA capture beads and PE detection reagents

TRUSTED_SUSTAINABILITY

Specifications

Buffer Aqueous Buffered Solution containing BSA and ProClin™ 150
Includes Manual
Product Type Cytometric Ab Control Standards
Formulation Mixture of individually lyophilized recombinant proteins made up of recombinant mouse interleukin (IL)-2, IL-4, IL-5, Interferon-γ (IFN-γ) and tumor necrosis factor (TNF)
For Use With (Application) Flow cytometry
pH 7.2
Regulatory Status RUO
Storage Requirements 4°C
Form Lyophilized
Quantity 1 Vial
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis