Learn More
BD Difco™ Adjuvants
Used to prepare antigen-adjuvant emulsions. BD™ Difco™ Adjuvants are used in immunological studies with laboratory animals. (Not intended for human or therapeutic use.)
Brand:  BD 231141
Code : TP
Additional Details : Weight : 0.02970kg
 View more versions of this product
Product Code. 10218823
  • £1010.00 / 600mg
Estimated Shipment: 22-08-2024
to see stock.
Alert:
Not intended for human or therapeutic use.
Description
Description
  • Adjuvants are mixed with aqueous antigen solution or suspension
  • Once injected, they provide a pool of emulsified antigen at the injection site and allow the antigen to be released over an extended period of time
  • With adjuvants, the immune response to antigens (especially soluble antigens) is enhanced and prolonged
  • Use Incomplete Freund adjuvant to avoid the use of acid-fast bacilli; however, the antibody response is not as marked as with the complete adjuvant
Specifications
Specifications
Store in cool, dry conditions in well sealed containers, away from oxidizing agents.
Mycobacterium tuberculosis H37Ra
6 x 100 mg
Killed M. tuberculosis H37 Ra for use in adjuvants
Immunological studies with laboratory animals