Learn More
BD Difco™ Adjuvants
Used to prepare antigen-adjuvant emulsions. BD™ Difco™ Adjuvants are used in immunological studies with laboratory animals. (Not intended for human or therapeutic use.)
Brand:  BD 263810
Additional Details : CAS Number : 8042-47-5 Weight : 0.13950kg
 View more versions of this product
Product Code. 11719062
  • £231.00 / 60mL
Estimated Shipment: 07-08-2024
to see stock.
Alert:
Not intended for human or therapeutic use.
Description
Description
  • Adjuvants are mixed with aqueous antigen solution or suspension
  • Once injected, they provide a pool of emulsified antigen at the injection site and allow the antigen to be released over an extended period of time
  • With adjuvants, the immune response to antigens (especially soluble antigens) is enhanced and prolonged
  • Use Incomplete Freund adjuvant to avoid the use of acid-fast bacilli; however, the antibody response is not as marked as with the complete adjuvant
Specifications
Specifications
Store in cool, dry conditions in well sealed containers, away from oxidizing agents.
Freund's Complete Adjuvant
6 x 10 mL
Suspension of 5mg M. butyricum in a mixture of paraffin oil and an emulsifying agent
Immunological studies with laboratory animals